Cytosin

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

Synonyms: 4-Amino-2-hydroxypyrimidin4-Amino-2-pyrimidinol

ACTIAS20122G100EA 0 CHF
ACTIAS20122G100 ACTIAS20122G500
Cytosin
Cytosin
Formel: C₄H₅N₃O
MW: 111,1 g/mol
Melting Pt: 313…316 °C
Dichte: 0,99771 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006034
CAS-Nummer: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR