3-Amino-5-methylcapronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (+/-)-3-Amino-5-methylhexanoic acid, DL-beta-Homoleucine, 3-Amino-5-methylhexanoic acid

AS33790G1 AS33790G5
ACTIAS33790G1EA 0 CHF
ACTIAS33790G1 ACTIAS33790G5
3-Amino-5-methylcapronsäure
3-Amino-5-methylcapronsäure
Formel: C₇H₁₅NO₂
Melting Pt: 227 °C
MDL Number: MFCD01863281
CAS-Nummer: 3653-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR