2-Ethoxy-4-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS34057G1 AS34057G10 AS34057G5
ACTIAS34057G1EA 0 CHF
ACTIAS34057G1 ACTIAS34057G10 ACTIAS34057G5
2-Ethoxy-4-fluorphenylboronsäure
2-Ethoxy-4-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
MW: 183.97 g/mol
MDL Number: MFCD03095001
CAS-Nummer: 480438-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR