2-Brom-4-fluorbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-4-fluorobenzyl Alcohol, (2-Bromo-4-fluorophenyl)methanol

AS25266G100 AS25266G25
ACTIAS25266G100EA 0 CHF
ACTIAS25266G100 ACTIAS25266G25
2-Brom-4-fluorbenzylalkohol
2-Brom-4-fluorbenzylalkohol
Formel: C₇H₆BrFO
MDL Number: MFCD00672925
CAS-Nummer: 229027-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR