4-Brom-2,6-difluorbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Brom-2,6-difluorphenyl)methanol

AS34945G1 AS34945G5
ACTIAS34945G1EA 0 CHF
ACTIAS34945G1 ACTIAS34945G5
4-Brom-2,6-difluorbenzylalkohol
4-Brom-2,6-difluorbenzylalkohol
Formel: C₇H₅BrF₂O
MW: 223.02
MDL Number: MFCD03094461
CAS-Nummer: 162744-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR