3-Brom-4-hydroxybenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (3-Bromo-4-hydroxyphenyl)methanol, 2-Bromo-4-(hydroxymethyl)phenol

AS34256G1 AS34256G5
ACTIAS34256G1EA 0 CHF
ACTIAS34256G1 ACTIAS34256G5
3-Brom-4-hydroxybenzylalkohol
3-Brom-4-hydroxybenzylalkohol
Formel: C₇H₇BrO₂
MDL Number: MFCD00016869
CAS-Nummer: 29922-56-3
EINECS: 249-957-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR