2,4,6-Trichlorbenzylalkohol
Lieferant: ACTIVATE SCIENTIFIC
AS35160G1
AS35160G5
ACTIAS35160G1EA
0
CHF
ACTIAS35160G1
ACTIAS35160G5
2,4,6-Trichlorbenzylalkohol
2,4,6-Trichlorbenzylalkohol
Formel:
C₇H₅Cl₃O |
MDL Number:
MFCD03093891 CAS-Nummer: 217479-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...