1-(4-Nitrophenyl)piperidin
Lieferant: ACTIVATE SCIENTIFIC
AS21801G100
AS21801G25
AS21801G5
ACTIAS21801G100EA
369
CHF
ACTIAS21801G100
ACTIAS21801G25
ACTIAS21801G5
1-(4-Nitrophenyl)piperidin
1-(4-Nitrophenyl)piperidin
Formel:
C₁₁H₁₄N₂O₂ MW: 206,24 g/mol |
MDL Number:
MFCD00023662 CAS-Nummer: 6574-15-8 EINECS: 229-492-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...