Oxazol
Lieferant: ACTIVATE SCIENTIFIC
AS36310G1
AS36310G5
ACTIAS36310G1EA
0
CHF
ACTIAS36310G1
ACTIAS36310G5
Oxazol
Oxazole
Formel:
C₃H₃NO Siedepunkt: 68…69 °C (1013 hPa) Melting Pt: –87 °C Dichte: 1.08 g/cm³ (15 °C) |
MDL Number:
MFCD00009751 CAS-Nummer: 288-42-6 EINECS: 206-020-8 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...