7-Nitro-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

AS21071G25 AS21071G5
ACTIAS21071G25EA 161 CHF
ACTIAS21071G25 ACTIAS21071G5
7-Nitro-1H-indazol
7-Nitro-1H-indazole
Formel: C₇H₅N₃O₂
MW: 163,14 g/mol
Melting Pt: 190 °C
Dichte: 1,56 g/cm³ (293 °C)
MDL Number: MFCD00022789
CAS-Nummer: 2942-42-9
EINECS: 220-934-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR