6-Amino-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Aminoindazole, 1H-Indazol-6-amine, 6-Amino-1H-indazole, Indazol-6-ylamine, Indazol-6-amine, 1H-Indazol-6-ylamine

AS20594G100 AS20594G25
ACTIAS20594G100EA 0 CHF
ACTIAS20594G100 ACTIAS20594G25
6-Amino-1H-indazol
6-Amino-1H-indazole
Formel: C₇H₇N₃
Siedepunkt: 345 °C (1013 hPa)
Melting Pt: 210 °C
MDL Number: MFCD00005696
CAS-Nummer: 6967-12-0
EINECS: 230-177-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR