6-Aminobenzothiazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Amino-1,3-benzothiazole, Benzothiazol-6-ylamine

AS30619G1 AS30619G5
ACTIAS30619G1EA 0 CHF
ACTIAS30619G1 ACTIAS30619G5
6-Aminobenzothiazol
6-Aminobenzothiazole
Formel: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD00015461
CAS-Nummer: 533-30-2
EINECS: 208-559-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR