5-Methylthiazol

Lieferant: ACTIVATE SCIENTIFIC

AS35091G1 AS35091G10 AS35091G5
ACTIAS35091G1EA 0 CHF
ACTIAS35091G1 ACTIAS35091G10 ACTIAS35091G5
5-Methylthiazol
5-Methylthiazole
Formel: C₄H₅NS
Siedepunkt: 139 °C (1013 hPa)
Melting Pt: –40.4 °C
Dichte: 1,127 g/cm³ (20 °C)
MDL Number: MFCD00005335
CAS-Nummer: 3581-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR