5-Fluor-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
AS50656G1
AS50656G5
ACTIAS50656G1EA
0
CHF
ACTIAS50656G1
ACTIAS50656G5
5-Fluor-1H-indazol
5-Fluoro-1H-indazole
Formel:
C₇H₅FN₂ |
MDL Number:
MFCD04972877 CAS-Nummer: 348-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...