5-Brom-2-fluorpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS50116G1 AS50116G5
ACTIAS50116G1EA 0 CHF
ACTIAS50116G1 ACTIAS50116G5
5-Brom-2-fluorpyrimidin
5-Bromo-2-fluoropyrimidine
Formel: C₄H₂BrFN₂
MDL Number: MFCD07787364
CAS-Nummer: 62802-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR