5-Benzyloxyindol

Lieferant: ACTIVATE SCIENTIFIC

AS50143G100 AS50143G5
ACTIAS50143G100EA 0 CHF
ACTIAS50143G100 ACTIAS50143G5
5-Benzyloxyindol
5-Benzyloxyindol
Formel: C₁₅H₁₃NO
Siedepunkt: 165 °C (0,1 mmHg)
Melting Pt: 107 °C
MDL Number: MFCD00005676
CAS-Nummer: 1215-59-4
EINECS: 214-930-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR