2-(4-Methyl-1,3-thiazol-5-yl)ethanol
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
5-(2-Hydroxyethyl)-4-methylthiazol
, 4-Methyl-5-thiazoleethanol
, 4-Methyl-5-(2-hydroxyethyl)thiazol
ACTIAS40464G100EA
0
CHF
ACTIAS40464G100
2-(4-Methyl-1,3-thiazol-5-yl)ethanol
2-(4-Methylthiazol-5-yl)ethanol
Formel:
C₆H₉NOS MW: 143,21 g/mol Siedepunkt: 135 °C (1013 hPa) Dichte: 1,199 g/cm³ (20 °C) |
MDL Number:
MFCD00005339 CAS-Nummer: 137-00-8 EINECS: 205-272-6 Merck Index: 12,06208 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...