5-(2-Bromphenyl)-1H-tetrazol

Lieferant: ACTIVATE SCIENTIFIC

AS23654G25 AS23654G5
ACTIAS23654G25EA 0 CHF
ACTIAS23654G25 ACTIAS23654G5
5-(2-Bromphenyl)-1H-tetrazol
5-(2-Bromphenyl)-1H-tetrazol
Formel: C₇H₅BrN₄
Melting Pt: 183…184 °C
MDL Number: MFCD00151799
CAS-Nummer: 73096-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR