4-Chlor-3-(trifluormethyl)benzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS40423G1
AS40423G5
ACTIAS40423G1EA
0
CHF
ACTIAS40423G1
ACTIAS40423G5
4-Chlor-3-(trifluormethyl)benzaldehyd
4-Chloro-3-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄ClF₃O |
MDL Number:
MFCD00040722 CAS-Nummer: 34328-46-6 EINECS: 251-940-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...