4-Amino-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
AS1820G1
AS1820G5
ACTIAS1820G1EA
0
CHF
ACTIAS1820G1
ACTIAS1820G5
4-Amino-1H-indazol
4-Amino-1H-indazole
Formel:
C₇H₇N₃ |
MDL Number:
MFCD03305710 CAS-Nummer: 41748-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...