4-Amino-6-chlorpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Chloropyrimidin-4-ylamine, 6-Chloropyrimidin-4-amine, 6-Chloro-4-pyrimidinamine, 6-Chlor-4-pyrimidinamin

AS50763G100 AS50763G5
ACTIAS50763G100EA 0 CHF
ACTIAS50763G100 ACTIAS50763G5
4-Amino-6-chlorpyrimidin
4-Amino-6-chloropyrimidine
Formel: C₄H₄ClN₃
MW: 129,55 g/mol
MDL Number: MFCD00053576
CAS-Nummer: 5305-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR