4-Amino-3-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Iodopyridin-4-amine, 3-Iodo-4-pyridylamine

AS50653G1 AS50653G5
ACTIAS50653G1EA 0 CHF
ACTIAS50653G1 ACTIAS50653G5
4-Amino-3-iodpyridin
4-Amino-3-iodopyridine
Formel: C₅H₅IN₂
MW: 220,01 g/mol
MDL Number: MFCD02663853
CAS-Nummer: 88511-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR