4-Amino-3-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50327G1 AS50327G5
ACTIAS50327G1EA 0 CHF
ACTIAS50327G1 ACTIAS50327G5
4-Amino-3-fluorpyridin
4-Amino-3-fluoropyridine
Formel: C₅H₅FN₂
MW: 112,11 g/mol
MDL Number: MFCD00234072
CAS-Nummer: 2247-88-3
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR