4,6-Dimethylpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS32272G1 AS32272G5
ACTIAS32272G1EA 0 CHF
ACTIAS32272G1 ACTIAS32272G5
4,6-Dimethylpyrimidin
4,6-Dimethylpyrimidine
Formel: C₆H₈N₂
Siedepunkt: 154 °C (1013 hPa)
Melting Pt: 25 °C
Dichte: 0,98 g/cm³ (20 °C)
MDL Number: MFCD00006116
CAS-Nummer: 1558-17-4
EINECS: 216-314-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR