3-Nitrophthalimid

Lieferant: ACTIVATE SCIENTIFIC

AS20686G100 AS20686G25
ACTIAS20686G100EA 64 CHF
ACTIAS20686G100 ACTIAS20686G25
3-Nitrophthalimid
3-Nitrophthalimid
Formel: C₈H₄N₂O₄
MW: 192,13 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00041852
CAS-Nummer: 603-62-3
EINECS: 210-051-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR