3-Formylbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde

AS34269G1 AS34269G10 AS34269G5
ACTIAS34269G1EA 0 CHF
ACTIAS34269G1 ACTIAS34269G10 ACTIAS34269G5
3-Formylbenzoesäure
3-Formylbenzoesäure
Formel: C₈H₆O₃
MW: 150,13 g/mol
Siedepunkt: 306 °C (1013 hPa)
Melting Pt: 175 °C
MDL Number: MFCD00039575
CAS-Nummer: 619-21-6
EINECS: 210-585-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR