3-Chlor-1H-indazol
Lieferant: ACTIVATE SCIENTIFIC
AS31231G1
AS31231G5
ACTIAS31231G1EA
0
CHF
ACTIAS31231G1
ACTIAS31231G5
3-Chlor-1H-indazol
3-Chloro-1H-indazole
Formel:
C₇H₅ClN₂ Siedepunkt: 310 °C (1013 hPa) Melting Pt: 147…148 °C |
MDL Number:
MFCD00067527 CAS-Nummer: 29110-74-5 EINECS: 249-444-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...