3-Chlorisonicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

AS30574G1 AS30574G5
ACTIAS30574G1EA 0 CHF
ACTIAS30574G1 ACTIAS30574G5
3-Chlorisonicotinonitril
3-Chlorisonicotinonitril
Formel: C₆H₃ClN₂
MW: 138,56 g/mol
MDL Number: MFCD05663706
CAS-Nummer: 68325-15-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR