3-Amino-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1H-Indazol-3-amine, Indazol-3-amine, 1H-Indazol-3-ylamine, Indazol-3-ylamine, 3-Amino-1H-indazole

AS22563G1 AS22563G25 AS22563G5
ACTIAS22563G1EA 0 CHF
ACTIAS22563G1 ACTIAS22563G25 ACTIAS22563G5
3-Amino-1H-indazol
3-Amino-1H-indazole
Formel: C₇H₇N₃
MW: 133,15 g/mol
MDL Number: MFCD00182045
CAS-Nummer: 874-05-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR