5-Amino-2-fluorpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Fluoro-5-pyridylamine, 2-Fluoropyridin-5-amine
AS31192G1
AS31192G5
ACTIAS31192G1EA
0
CHF
ACTIAS31192G1
ACTIAS31192G5
5-Amino-2-fluorpyridin
5-Amino-2-fluoropyridine
Formel:
C₅H₅FN₂ MW: 112,11 g/mol |
MDL Number:
MFCD01632180 CAS-Nummer: 1827-27-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...