3-Aminophthalimid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Aminoisoindoline-1,3-dione

AS20690G1 AS20690G25 AS20690G5
ACTIAS20690G1EA 21 CHF
ACTIAS20690G1 ACTIAS20690G25 ACTIAS20690G5
3-Aminophthalimid
3-Aminophthalimid
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
Siedepunkt: 447 °C (1013 hPa)
Melting Pt: 270…271 °C
MDL Number: MFCD00041853
CAS-Nummer: 2518-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR