2-Desoxy-D(-)-ribose
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
2-Desoxy-D(-)-erythropentose
, 2-Desoxy-D(-)-arabinose
, Thyminose
ACTIAS31093G100EA
0
CHF
ACTIAS31093G100
2-Desoxy-D(-)-ribose
2-Desoxy-D(-)-ribose
Formel:
C₅H₁₀O₄ MW: 134,13 g/mol Siedepunkt: 292 °C (1013 hPa) Melting Pt: 95…97 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00135904 CAS-Nummer: 533-67-5 EINECS: 208-573-0 Merck Index: 13,02925 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...