5-Fluor-2-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoro-2-pyridinecarbonitrile, 2-Cyano-5-fluoropyridine

AS40226G1 AS40226G5
ACTIAS40226G1EA 0 CHF
ACTIAS40226G1 ACTIAS40226G5
5-Fluor-2-pyridincarbonitril
5-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
MW: 122,1 g/mol
MDL Number: MFCD08741371
CAS-Nummer: 327056-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR