3-Iod-2-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS31864G1 AS31864G5
ACTIAS31864G1EA 0 CHF
ACTIAS31864G1 ACTIAS31864G5
3-Iod-2-pyridincarbonitril
3-Iod-2-pyridincarbonitril
Formel: C₆H₃IN₂
MDL Number: MFCD09037451
CAS-Nummer: 827616-52-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR