2-Chlorbenzoxazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-1,3-benzoxazole

AS23173G25 AS23173G100
ACTIAS23173G25EA 0 CHF
ACTIAS23173G25 ACTIAS23173G100
2-Chlorbenzoxazol
2-Chlorobenzoxazole
Formel: C₇H₄ClNO
Siedepunkt: 202 °C (1013 hPa)
Melting Pt: 45 °C
Dichte: 1.3453 g/cm³ (18 °C)
MDL Number: MFCD00005766
CAS-Nummer: 615-18-9
EINECS: 210-414-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR