2-Chlor-6-pyridincarbonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Chloropicolinonitrile, 6-Chloropyridine-2-carbonitrile, 2-Chloro-6-cyanopyridine
AS7206G100
AS7206G5
ACTIAS7206G100EA
0
CHF
ACTIAS7206G100
ACTIAS7206G5
2-Chlor-6-pyridincarbonitril
2-Chlor-6-pyridincarbonitril
Formel:
C₆H₃ClN₂ MW: 138,56 g/mol |
MDL Number:
MFCD00274527 CAS-Nummer: 33252-29-8 EINECS: 251-429-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...