6-Chlornicotinonitril
Lieferant: ACTIVATE SCIENTIFIC
AS4006G100
AS4006G5
ACTIAS4006G100EA
0
CHF
ACTIAS4006G100
ACTIAS4006G5
6-Chlornicotinonitril
6-Chlornicotinonitril
Formel:
C₆H₃ClN₂ |
MDL Number:
MFCD00084941 CAS-Nummer: 33252-28-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...