2-Brom-5-(trifluormethyl)benzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS31127G1
AS31127G5
ACTIAS31127G1EA
0
CHF
ACTIAS31127G1
ACTIAS31127G5
2-Brom-5-(trifluormethyl)benzaldehyd
2-Bromo-5-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄BrF₃O MW: 253.02 g/mol |
MDL Number:
MFCD04973760 CAS-Nummer: 102684-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...