2-Amino-6-methoxybenzothiazol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS31472G100EA
0
CHF
ACTIAS31472G100
2-Amino-6-methoxybenzothiazol
2-Amino-6-methoxybenzothiazole
Formel:
C₈H₈N₂OS MW: 180,23 g/mol Siedepunkt: 348 °C (1013 hPa) Melting Pt: 160 °C |
MDL Number:
MFCD00005787 CAS-Nummer: 1747-60-0 EINECS: 217-130-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...