2-Amino-6-brombenzothiazol

Lieferant: ACTIVATE SCIENTIFIC

AS31549G1 AS31549G5
ACTIAS31549G1EA 0 CHF
ACTIAS31549G1 ACTIAS31549G5
2-Amino-6-brombenzothiazol
2-Amino-6-bromobenzothiazole
Formel: C₇H₅BrN₂S
MW: 229,1 g/mol
MDL Number: MFCD00152229
CAS-Nummer: 15864-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR