2-Amino-5-methylthiazol

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Methylthiazol-2-ylamin

ACTIAS40161G100EA 0 CHF
ACTIAS40161G100
2-Amino-5-methylthiazol
2-Amino-5-methylthiazole
Formel: C₄H₆N₂S
MW: 114,17 g/mol
Siedepunkt: 222 °C (1013 hPa)
Melting Pt: 96 °C
MDL Number: MFCD00078317
CAS-Nummer: 7305-71-7
EINECS: 423-800-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR