2-Aminoisonicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Aminopyridine-4-carbonitrile, 2-Amino-4-cyanopyridine

AS4042G1 AS4042G25 AS4042G5
ACTIAS4042G1EA 0 CHF
ACTIAS4042G1 ACTIAS4042G25 ACTIAS4042G5
2-Aminoisonicotinonitril
2-Aminoisonicotinonitril
Formel: C₆H₅N₃
MW: 119.13 g/mol
MDL Number: MFCD03791310
CAS-Nummer: 42182-27-4
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR