2-Amino-4-(trifluormethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS21722G1 AS21722G5
ACTIAS21722G1EA 19 CHF
ACTIAS21722G1 ACTIAS21722G5
2-Amino-4-(trifluormethyl)pyridin
2-Amino-4-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
MW: 162,11 g/mol
MDL Number: MFCD00077455
CAS-Nummer: 106447-97-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR