2-Amino-3-chlorpyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS50650G5EA 0 CHF
ACTIAS50650G1 ACTIAS50650G5
2-Amino-3-chlorpyridin
2-Amino-3-chloropyridine
Formel: C₅H₅ClN₂
MDL Number: MFCD03541052
CAS-Nummer: 39620-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR