2,6-Dichlorbenzothiazol

Lieferant: ACTIVATE SCIENTIFIC

AS33304G1 AS33304G5
ACTIAS33304G1EA 0 CHF
ACTIAS33304G1 ACTIAS33304G5
2,6-Dichlorbenzothiazol
2,6-Dichlorobenzothiazole
Formel: C₇H₃Cl₂NS
MW: 204,08 g/mol
MDL Number: MFCD00044101
CAS-Nummer: 3622-23-9
EINECS: 222-819-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR