2,6-Dichlor-4-(trifluormethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS31947G5EA 0 CHF
ACTIAS31947G5 ACTIAS31947G1
2,6-Dichlor-4-(trifluormethyl)pyridin
2,6-Dichloro-4-(trifluoromethyl)pyridine
Formel: C₆H₂Cl₂F₃N
MW: 215,99 g/mol
MDL Number: MFCD00042246
CAS-Nummer: 39890-98-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR