2,5-Dibromthiazol
Lieferant: ACTIVATE SCIENTIFIC
AS50264G1
AS50264G5
ACTIAS50264G1EA
0
CHF
ACTIAS50264G1
ACTIAS50264G5
2,5-Dibromthiazol
2,5-Dibromothiazole
Formel:
C₃HBr₂NS |
MDL Number:
MFCD00016891 CAS-Nummer: 4175-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...