2,5-Dibrombenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS31352G1
AS31352G5
ACTIAS31352G1EA
0
CHF
ACTIAS31352G1
ACTIAS31352G5
2,5-Dibrombenzaldehyd
2,5-Dibromobenzaldehyde
Formel:
C₇H₄Br₂O MW: 263,92 g/mol |
MDL Number:
MFCD00462864 CAS-Nummer: 74553-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...