5-Fluoruracil
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Gefahr
|
Synonyms:
2,4-Dihydroxy-5-fluorpyrimidin
, Fluoruracil
, 5-FU
ACTIAS21439G100EA
90
CHF
ACTIAS21439G100
5-Fluoruracil
5-Fluoruracil
Formel:
C₄H₃FN₂O₂ MW: 130,08 g/mol Siedepunkt: 347 °C (1013 hPa) Melting Pt: 282…283 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00006018 CAS-Nummer: 51-21-8 EINECS: 200-085-6 UN: 2811 ADR: 6.1,III Merck Index: 13,04208 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...