1-(2-Pyridyl)piperazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(1-Piperazinyl)pyridine

AS23860G100 AS23860G25
ACTIAS23860G100EA 0 CHF
ACTIAS23860G100 ACTIAS23860G25
1-(2-Pyridyl)piperazin
1-(2-Pyridyl)piperazin
Formel: C₉H₁₃N₃
MW: 163.22 g/mol
Siedepunkt: 293
Melting Pt: 85…87 °C
Dichte: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00006216
CAS-Nummer: 34803-66-2
EINECS: 252-220-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR